Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Citric acid monohydrate
Synonym(s):
Citric acid monohydrate
Linear Formula:
HOC(COOH)(CH
2
COOH)
2
· H
2
O
CAS Number:
5949-29-1
Molecular Weight:
210.14
EC Number:
201-069-1
Products
Product Category
acids (4)
antioxidants (3)
building blocks (3)
organic acids (3)
flavorings (2)
buffers (1)
Show More
Brand
SAFC (3)
Sigma-Aldrich (7)
Supelco (4)
Color
Feature
Formula Weight
Physical Form
pK Value
Product Line
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Special Grade
ACS reagent (1)
Puriss P.A. (1)
certified reference material (1)
pharmaceutical primary standard (1)
pharmaceutical secondary standard (1)
Filter Products
Showing 1-15 of 15
Compare
Product Number
Description
Pricing
1.00244
Citric acid monohydrate,
for analysis EMSURE
®
ACS,ISO,Reag. Ph Eur
Expand
View Pricing
C1909
Citric acid monohydrate,
ACS reagent, ≥99.0%
Expand
View Pricing
1.37003
Citric acid monohydrate,
crystalline, EMPROVE
®
EXPERT, Ph. Eur., BP, ChP, JP, USP
Expand
View Pricing
1.00243
Citric acid monohydrate,
powder, EMPROVE
®
ESSENTIAL, Ph. Eur., BP, ChP, JP, USP, FCC, E 330
Expand
View Pricing
1.00242
Citric acid monohydrate,
crystalline, EMPROVE
®
ESSENTIAL, Ph. Eur., BP, ChP, USP, FCC, E 330
Expand
View Pricing
C7129
Citric acid monohydrate,
reagent grade, ≥98% (GC/titration)
Expand
View Pricing
33114
Citric acid monohydrate,
puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., buffer substance, 99.5-102%
Expand
View Pricing
1.93411
Citric acid monohydrate,
EMPLURA
®
Expand
View Pricing
1.93011
Citric acid monohydrate,
for analysis EMPARTA
®
ACS
Expand
View Pricing
05-4930
Citric acid monohydrate,
JIS special grade, ≥99.5%
Expand
View Pricing
PHR2903
Citric acid monohydrate,
pharmaceutical secondary standard, certified reference material
Expand
View Pricing
C0706
Citric acid monohydrate,
BioXtra, ≥99.5%
Expand
View Pricing
05-4940
Citric acid monohydrate,
SAJ first grade, ≥99.5%
Expand
View Pricing
C2219000
Citric acid monohydrate,
European Pharmacopoeia (EP) Reference Standard
Expand
View Pricing
05-4950
Citric acid monohydrate,
≥99.5%, suitable for amino acid analysis
Expand
View Pricing