Skip to Content
Merck
Chloroethane solution 2.0 M in tert-butyl methyl ether, anhydrous

Chloroethane solution

Chloroethane solution 2.0 M in tert-butyl methyl ether, anhydrous
Synonym(s):
Chloroethane solution, Ethyl chloride
Empirical Formula (Hill Notation):
C2H5Cl
CAS Number:
Molecular Weight:
64.51

Products

Product Category
Brand
Formula Weight
Physical Form
Product Line
Format
Application
Markush Class
Markush Group
Package Size
Reference Material Type
Special Grade
Technique