
Chiralyst P280

Synonym(s):
Dichloro(1,5-cyclooctadiene)ruthenium(II), polymer, (1,5-Cyclooctadiene)ruthenium(II) chloride, Ruthenium(II) chloride 1,5-cyclooctadiene complex
Linear Formula:
[Ru(COD)Cl2]n
CAS Number:
Molecular Weight:
280.16 (as monomer)
Products
Brand
Formula Weight
Physical Form
Purity
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing