Chenodeoxycholic acid
Synonym(s):
3α,7α-Dihydroxy-5β-cholanic acid, 5β-Cholanic acid-3α,7α-diol, Chenodiol
Empirical Formula (Hill Notation):
C24H40O4
CAS Number:
Molecular Weight:
392.57
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Format
Quality Segment
Functional Group
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3