
Celite® 545 AW Inorganic Adsorbent

Synonym(s):
Diatomaceous earth, flux-calcined
CAS Number:
Products
Product Category
Brand
Physical Form
Agency
Separation Technique
Quality Segment
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing