
Cefazolin sodium salt

Empirical Formula (Hill Notation):
C14H13N8NaO4S3
CAS Number:
Molecular Weight:
476.49
Products
Product Category
Brand
Feature
Formula Weight
Physical Form
Purity
Activity Spectrum
Agency
Quality Segment
Application
Mode of Action
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing