Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Carbotrap
®
T420 Thermal Desorption Tube
Synonym(s):
Carbotrap
®
T420
Products
Brand
Supelco (2)
Sustainability
Feature
Product Line
Agency
Length (mm)
OD (mm)
Quality Segment
Package Size
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
28687-U
Carbotrap
®
T420 Thermal Desorption Tube,
316 stainless steel, O.D. x I.D. x L 1/4 in. (6.35 mm) × 3 1/2 in. (89 mm), preconditioned, pkg of 10 tubes
Expand
View Pricing
28689-U
Carbotrap
®
T420 Thermal Desorption Tube,
glass, O.D. x I.D. x L 1/4 in. (6.35 mm) × 3 1/2 in. (89 mm), fritted, preconditioned, pkg of 10 tubes
Expand
View Pricing