Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Carbon dioxide in nitrogen
Products
Manufacturer
Format
Quality Segment
Application
Reference Material Type
Special Grade
certified reference material (4)
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
NIST2745
Carbon dioxide in nitrogen,
NIST SRM 2745, nominal 16% mol/mol
Expand
View Pricing
NIST2620A
Carbon dioxide in nitrogen,
NIST SRM 2620a, nominal 1 % mol/mol
Expand
View Pricing
NIST2621A
Carbon dioxide in nitrogen,
NIST
®
SRM
®
2621a, nominal 1.5% mol/mol
Expand
View Pricing
NIST2624A
Carbon dioxide in nitrogen,
NIST
®
SRM
®
2624a, nominal 3% mol/mol
Expand
View Pricing
NIST2619A
Carbon dioxide in nitrogen,
NIST
®
SRM
®
2619a, nominal 0.5% mol/mol
Expand
View Pricing