
Campesterol

Synonym(s):
24α-Methyl-5-cholesten-3β-ol, 24(R)-Ergost-5-en-3β-ol
Empirical Formula (Hill Notation):
C28H48O
CAS Number:
Molecular Weight:
400.68
Products
Product Category
Brand
Biological Source
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing