Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Cameo
™
Syringe Filter
Products
Product Category
syringe filters (8)
Manufacturer
Diameter (mm)
Housing Material
Membrane Material
Pore size (µm)
Sterility
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
Z741966
Cameo
™
Syringe Filter,
PTFE membrane, pore size 0.22 μm, diam. 30 mm
Expand
View Pricing
Z741961
Cameo
™
Syringe Filter,
PVDF membrane, pore size 0.22 μm, diam. 30 mm
Expand
View Pricing
Z741962
Cameo
™
Syringe Filter,
PVDF membrane, pore size 0.45 μm, diam. 17 mm
Expand
View Pricing
Z741960
Cameo
™
Syringe Filter,
polypropylene membrane, pore size 0.45 μm, diam. 30 mm
Expand
View Pricing
Z741951
Cameo
™
Syringe Filter,
polyethersulfone membrane, pore size 0.45 μm, diam. 17 mm
Expand
View Pricing
Z741954
Cameo
™
Syringe Filter,
polyethersulfone membrane, pore size 0.45 μm, diam. 30 mm
Expand
View Pricing
Z741967
Cameo
™
Syringe Filter,
PTFE membrane, pore size 0.45 μm, diam. 17 mm
Expand
View Pricing
Z741963
Cameo
™
Syringe Filter,
PVDF membrane, pore size 0.45 μm, diam. 30 mm
Expand
View Pricing