
Calcium L-lactate hydrate

Synonym(s):
(S)-(+)-2-Hydroxypropanoic acid hemicalcium salt, L-Lactic acid hemicalcium salt, Sarcolactic acid hemicalcium salt
Linear Formula:
[CH3CH(OH)COO]2Ca · xH2O
Products
Product Category
Brand
Biological Source
Formula Weight
Physical Form
Product Line
Purity
Markush Class
Markush Group
Optical Isomer
Special Grade
Technique
Showing 1-3 of 3