Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Calcium chloride solution
Empirical Formula (Hill Notation):
CaCl
2
CAS Number:
10043-52-4
Molecular Weight:
110.98
Products
Product Category
salts (3)
Brand
Sigma-Aldrich (3)
Formula Weight
pH Value
Physical Form
Product Line
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
21115
Calcium chloride solution,
BioUltra, Molecular Biology, ~1 M in H
2
O
Expand
View Pricing
05-0535
Calcium chloride solution,
0.025 M
Expand
View Pricing
05-0604
Calcium chloride solution,
3.2 mM
Expand
View Pricing