
Bromoform-13C

Synonym(s):
Tribromomethane-13C
Linear Formula:
13CHBr3
CAS Number:
Molecular Weight:
253.72
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Isotope
Showing 1-1 of 1
Compare
Product Number
Description
Pricing