Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Bottle Tag, For Gl 45 Duran
®
Media Bottles Bottle Tag
Products
Product Category
bottles (8)
Bottle material
Color
Vial Bottle Material
Vial Bottle Color
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
DWK292432859
Bottle Tag, For Gl 45 Duran
®
Media Bottles Bottle Tag,
blue silicone bottle
Expand
View Pricing
DWK292432818
Bottle Tag, For Gl 45 Duran
®
Media Bottles Bottle Tag,
red silicone bottle
Expand
View Pricing
DWK292432883
Bottle Tag, For Gl 45 Duran
®
Media Bottles Bottle Tag,
white silicone bottle
Expand
View Pricing
DWK292432834
Bottle Tag, For Gl 45 Duran
®
Media Bottles Bottle Tag,
yellow silicone bottle
Expand
View Pricing
DWK292432875
Bottle Tag, For Gl 45 Duran
®
Media Bottles Bottle Tag,
black silicone bottle
Expand
View Pricing
DWK292432842
Bottle Tag, For Gl 45 Duran
®
Media Bottles Bottle Tag,
green silicone bottle
Expand
View Pricing
DWK292432826
Bottle Tag, For Gl 45 Duran
®
Media Bottles Bottle Tag,
orange silicone bottle
Expand
View Pricing
DWK292432867
Bottle Tag, For Gl 45 Duran
®
Media Bottles Bottle Tag,
purple silicone bottle
Expand
View Pricing