Skip to Content
Merck
Borax Solution 5 g/dL in deionized water

Borax Solution

Borax Solution 5 g/dL in deionized water
Synonym(s):
Borax solution, Sodium borate solution

Products

Product Category
Brand
Physical Form
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing