Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
BIOshell A160 Peptide PCS-C18, 2.7 µm HPLC Column
Products
Brand
Supelco (15)
ID (mm)
Length (mm)
Quality Segment
Package Size
Filter Products
Showing 1-15 of 15
Compare
Product Number
Description
Pricing
50668-U
BIOshell A160 Peptide PCS-C18, 2.7 µm HPLC Column,
column L × I.D. 150 mm × 4.6 mm, pk of 1 ea
Expand
View Pricing
50659-U
BIOshell A160 Peptide PCS-C18, 2.7 µm HPLC Column,
column L × I.D. 150 mm × 2.1 mm, pk of 1 ea
Expand
View Pricing
50660-U
BIOshell A160 Peptide PCS-C18, 2.7 µm HPLC Column,
column L × I.D. 50 mm × 3.0 mm, pk of 1 ea
Expand
View Pricing
50657-U
BIOshell A160 Peptide PCS-C18, 2.7 µm HPLC Column,
column L × I.D. 100 mm × 2.1 mm, pk of 1 ea
Expand
View Pricing
50656-U
BIOshell A160 Peptide PCS-C18, 2.7 µm HPLC Column,
column L × I.D. 50 mm × 2.1 mm, pk of 1 ea
Expand
View Pricing
50653-U
BIOshell A160 Peptide PCS-C18, 2.7 µm HPLC Column,
column L × I.D. 50 mm × 1.5 mm, pk of 1 ea
Expand
View Pricing
50654-U
BIOshell A160 Peptide PCS-C18, 2.7 µm HPLC Column,
column L × I.D. 100 mm × 1.5 mm, pk of 1 ea
Expand
View Pricing
50680-U
BIOshell A160 Peptide PCS-C18, 2.7 µm HPLC Column,
L × I.D. 5 mm × 4.6 mm, pk of 3 ea, guard columns
Expand
View Pricing
50675-U
BIOshell A160 Peptide PCS-C18, 2.7 µm HPLC Column,
L × I.D. 5 mm × 3.0 mm, pk of 3 ea, guard columns
Expand
View Pricing
50655-U
BIOshell A160 Peptide PCS-C18, 2.7 µm HPLC Column,
column L × I.D. 150 mm × 1.5 mm, pk of 1 ea
Expand
View Pricing
50664-U
BIOshell A160 Peptide PCS-C18, 2.7 µm HPLC Column,
column L × I.D. 50 mm × 4.6 mm, pk of 1 ea
Expand
View Pricing
50663-U
BIOshell A160 Peptide PCS-C18, 2.7 µm HPLC Column,
column L × I.D. 150 mm × 3.0 mm, pk of 1 ea
Expand
View Pricing
50665-U
BIOshell A160 Peptide PCS-C18, 2.7 µm HPLC Column,
column L × I.D. 100 mm × 4.6 mm, pk of 1 ea
Expand
View Pricing
50661-U
BIOshell A160 Peptide PCS-C18, 2.7 µm HPLC Column,
column L × I.D. 100 mm × 3.0 mm, pk of 1 ea
Expand
View Pricing
50671-U
BIOshell A160 Peptide PCS-C18, 2.7 µm HPLC Column,
L × I.D. 5 mm × 2.1 mm, pk of 3 ea, guard columns
Expand
View Pricing