Skip to Content
Merck
Benzyl benzoate SAJ first grade, ≥98.0%

Benzyl benzoate

Benzyl benzoate SAJ first grade, ≥98.0%
Synonym(s):
Benzyl benzoate, Benzoic acid benzyl ester
Linear Formula:
C6H5COOCH2C6H5
CAS Number:
Molecular Weight:
212.24

Products

Product Category
Brand
Sustainability
Boiling Point (°C)
Feature
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
API Family
Reference Material Type
Special Grade
Technique