Skip to Content
Merck
Bendiocarb PESTANAL®, analytical standard

Bendiocarb

Bendiocarb PESTANAL®, analytical standard
Empirical Formula (Hill Notation):
C11H13NO4
CAS Number:
Molecular Weight:
223.23
EC Number:

Products

Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing