Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
β-Alanine
Synonym(s):
β-Ala, 3-Aminopropionic acid
Linear Formula:
NH
2
CH
2
CH
2
COOH
CAS Number:
107-95-9
Molecular Weight:
89.09
EC Number:
203-536-5
Products
Product Category
amino acids (5)
Brand
Sigma-Aldrich (4)
Supelco (1)
Color
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Format
Analyte Suitability
Quality Segment
Application
Amino Acid
Reference Material Type
Special Grade
Puriss (1)
Puriss P.A. (1)
analytical standard (1)
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
146064
β-Alanine,
99%, suitable for ligand binding assays
Expand
View Pricing
A9920
β-Alanine,
BioReagent, suitable for cell culture, suitable for insect cell culture
Expand
View Pricing
05160
β-Alanine,
BioXtra, ≥99.0% (NT)
Expand
View Pricing
05159
β-Alanine,
BioUltra, ≥99.0% (NT)
Expand
View Pricing
41144
β-Alanine,
analytical standard
Expand
View Pricing