Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Autoclave disposal bags
Products
Product Category
bags (3)
Manufacturer
Height (mm)
Color
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
HS1002A
Autoclave disposal bags,
H × W × thickness 660 mm × 310 mm × 2.0 mm, transparent polypropylene (with blue biohazard labeling )
Expand
View Pricing
HS1002B
Autoclave disposal bags,
H × W × thickness 600 mm × 415 mm × 2.0 mm, transparent polypropylene (with blue biohazard labeling )
Expand
View Pricing
HS1002C
Autoclave disposal bags,
H × W × thickness 810 mm × 610 mm × 2.0 mm, transparent polypropylene (with blue biohazard labeling )
Expand
View Pricing