
ASTM® D6584 1,2,4-Butanetriol Solution

Synonym(s):
Biodiesel impurity, Butanetriol solution
CAS Number:
Products
Brand
Product Line
Agency
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing