
Arginine aspartate

Synonym(s):
L-Arginine L-aspartate salt, S(+)-2-Amino-5-[(aminoiminomethyl)amino]pentanoic acid monohydrochloride
Empirical Formula (Hill Notation):
C4H7NO4 · C6H14N4O2
CAS Number:
Molecular Weight:
307.30
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2