Anti-Vinculin antibody, Mouse monoclonal
Synonym(s):
Monoclonal Anti-Vinculin antibody produced in mouse, Anti-CMD1W, Anti-MVCL, Anti-VCL
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Antibody Form
Research Area
Conjugate
Application
Package Size
Post Translational Modification
Species Reactivity
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing