Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aniline
Synonym(s):
Aniline
Linear Formula:
C
6
H
5
NH
2
CAS Number:
62-53-3
Molecular Weight:
93.13
EC Number:
200-539-3
Products
Brand
Sigma-Aldrich (5)
Supelco (4)
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Special Grade
ACS reagent (1)
analytical standard (1)
certified reference material (1)
pharmaceutical primary standard (1)
pharmaceutical secondary standard (1)
Technique
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
8.22256
Aniline,
for synthesis
Expand
View Pricing
242284
Aniline,
ACS reagent, ≥99.5%
Expand
View Pricing
1.01261
Aniline,
for analysis EMSURE
®
Expand
View Pricing
1.94610
Aniline,
for analysis EMPARTA
®
ACS
Expand
View Pricing
132934
Aniline,
ReagentPlus
®
, 99%
Expand
View Pricing
51788
Aniline,
analytical standard
Expand
View Pricing
1036110
Aniline,
United States Pharmacopeia (USP) Reference Standard
Expand
View Pricing
01-5600
Aniline,
JIS special grade, ≥99.0%
Expand
View Pricing
PHR2346
Aniline,
Pharmaceutical Secondary Standard; Certified Reference Material
Expand
View Pricing
01-5610
Aniline,
SAJ first grade, ≥99.0%
Expand
View Pricing