![Amphotericin B solubilized powder, γ-irradiated, BioXtra, suitable for cell culture](/deepweb/assets/sigmaaldrich/product/structures/140/115/a0c927b2-145e-4d58-8265-4ecdb3f66477/640/a0c927b2-145e-4d58-8265-4ecdb3f66477.png)
Amphotericin B solubilized
![Amphotericin B solubilized powder, γ-irradiated, BioXtra, suitable for cell culture](/deepweb/assets/sigmaaldrich/product/structures/140/115/a0c927b2-145e-4d58-8265-4ecdb3f66477/640/a0c927b2-145e-4d58-8265-4ecdb3f66477.png)
Synonym(s):
Amphotericin B from Streptomyces sp., Fungizone
Empirical Formula (Hill Notation):
C47H73NO17
CAS Number:
Molecular Weight:
924.08
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Activity Spectrum
Quality Segment
Mode of Action
Sterilization Method
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing