Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ammonium sulfide solution
Linear Formula:
(NH
4
)
2
S
CAS Number:
12135-76-1
Molecular Weight:
68.14
Products
Product Category
salts (2)
aroma chemicals (1)
Brand
Sigma-Aldrich (3)
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Agency
Quality Segment
Application
Organoleptic Class
Special Grade
Halal (1)
Kosher (1)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
A1952
Ammonium sulfide solution,
20% in H
2
O
Expand
View Pricing
515809
Ammonium sulfide solution,
40-48 wt. % in H
2
O
Expand
View Pricing
W205303
Ammonium sulfide solution,
20 wt. % in H
2
O
Expand
View Pricing