Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ammonium peroxodisulfate
Synonym(s):
Ammonium peroxodisulfate
Linear Formula:
(NH
4
)
2
S
2
O
8
CAS Number:
7727-54-0
Molecular Weight:
228.20
Products
Product Category
salts (2)
Brand
Supelco (4)
Formula Weight
Physical Form
Product Line
Agency
Quality Segment
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
1.01201
Ammonium peroxodisulfate,
for analysis EMSURE
®
ACS,Reag. Ph Eur
Expand
View Pricing
1.01200
Ammonium peroxodisulfate,
EMPLURA
®
Expand
View Pricing
1.93228
Ammonium peroxodisulfate,
for analysis EMPARTA
®
ACS
Expand
View Pricing
1.93628
Ammonium peroxodisulfate,
EMPLURA
®
Expand
View Pricing