Skip to Content
Merck
Ammonium cerium(IV) nitrate JIS special grade, ≥95.0%

Ammonium cerium(IV) nitrate

Ammonium cerium(IV) nitrate JIS special grade, ≥95.0%
Synonym(s):
Ceric ammonium nitrate, Ammonium cerium(IV) nitrate
Linear Formula:
Ce(NH4)2(NO3)6
CAS Number:
Molecular Weight:
548.22
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Product Line
Purity
Agency
Quality Segment
Markush Class
Markush Group
Reagent Type
Special Grade