Skip to Content
Merck
Amberlite™ IRA402 chloride form chloride form

Amberlite IRA-402 Ion Exchange Resin

Amberlite™ IRA402 chloride form chloride form
Synonym(s):
Ion exchanger Amberlite IRA-402
CAS Number:

Products

Product Category
Brand
pH Value
Physical Form
Separation Technique
Quality Segment
Particle Size (µm)
Technique