Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aldrich
®
Graham condenser
Products
Brand
Sigma-Aldrich (7)
Height (mm)
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
Z531383
Aldrich
®
Graham condenser,
Overall H 200 mm, joint: ST/NS 24/40
Expand
View Pricing
Z531367
Aldrich
®
Graham condenser,
Overall H 100 mm, joint: ST/NS 14/20
Expand
View Pricing
Z531448
Aldrich
®
Graham condenser,
Overall H 400 mm, joint: ST/NS 24/40
Expand
View Pricing
Z531375
Aldrich
®
Graham condenser,
Overall H 200 mm, joint: ST/NS 14/20
Expand
View Pricing
Z531405
Aldrich
®
Graham condenser,
Overall H 300 mm, joint: ST/NS 14/20
Expand
View Pricing
Z531480
Aldrich
®
Graham condenser,
Overall H 600 mm, joint: ST/NS 24/40
Expand
View Pricing
Z531413
Aldrich
®
Graham condenser,
Overall H 300 mm, joint: ST/NS 24/40
Expand
View Pricing