
Ademetionine disulfate tosylate

Synonym(s):
S-Adenosyl-L-methionine disulfate tosylate
Empirical Formula (Hill Notation):
C22H34N6O16S4
CAS Number:
Molecular Weight:
766.80
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2