Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Adapter for Eppendorf
®
F-34-6-38 Rotor
Products
Product Category
adapters (9)
Manufacturer
Diameter (mm)
Package Size
Technique
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
ADF3463850-EP
Adapter for Eppendorf
®
F-34-6-38 Rotor,
holds 1 x 50 mL conical tube, pack of 2 ea
Expand
View Pricing
ADF346385-EP
Adapter for Eppendorf
®
F-34-6-38 Rotor,
holds 1 x 15 mL conical tube, pack of 2 ea
Expand
View Pricing
ADF3463827-EP
Adapter for Eppendorf
®
F-34-6-38 Rotor,
holds 2 x 7 - 15 mL round bottom-tubes, pack of 2 ea
Expand
View Pricing
ADF3463850R-EP
Adapter for Eppendorf
®
F-34-6-38 Rotor,
holds 1 x 50 mL round-bottom tube, pack of 2 ea
Expand
View Pricing
ADF346382-EP
Adapter for Eppendorf
®
F-34-6-38 Rotor,
holds 1 x 20 – 30 mL round-bottom tube, pack of 2 ea
Expand
View Pricing
ADF346381-EP
Adapter for Eppendorf
®
F-34-6-38 Rotor,
holds 4 x 1.5 - 2 mL reaction vessels, pack of 2 ea
Expand
View Pricing
EP5804777000
Adapter for Eppendorf
®
F-34-6-38 Rotor,
holds 1 Eppendorf Tube
®
5.0 mL, pack of 2
Expand
View Pricing
ADF346388-EP
Adapter for Eppendorf
®
F-34-6-38 Rotor,
holds 1 x 15 – 18 mL round-bottom tube, pack of 2 ea
Expand
View Pricing
ADAPF346382-EP
Adapter for Eppendorf
®
F-34-6-38 Rotor,
holds 3 round-bottom tubes 2.6-7 mL, pack of 2 ea
Expand
View Pricing