Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
ABETA 42 in CSF
Synonym(s):
Aβ1-42 in CSF, Amyloid β1-42 peptide in cerebrospinal fluid
Products
Product Category
peptides (3)
Manufacturer
Agency
Reference Material Type
Special Grade
certified reference material (3)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
ERMDA482IFCC
ABETA 42 in CSF,
ERM
®
, certified reference material, High Level
Expand
View Pricing
ERMDA480IFCC
ABETA 42 in CSF,
ERM
®
, certified reference material, Low Level
Expand
View Pricing
ERMDA481IFCC
ABETA 42 in CSF,
ERM
®
, certified reference material, Medium Level
Expand
View Pricing