Skip to Content
Merck
9,10-Anthracenedicarbonitrile 97%

9,10-Anthracenedicarbonitrile

9,10-Anthracenedicarbonitrile 97%
Synonym(s):
9,10-Dicyanoanthracene
Empirical Formula (Hill Notation):
C16H8N2
CAS Number:
Molecular Weight:
228.25

Products

Product Category
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing