
6,6-Dimethylfulvene

Synonym(s):
5-(1-Methylethylidene)-1,3-cyclopentadiene, 5-Isopropylidene-1,3-cyclopentadiene
Empirical Formula (Hill Notation):
C8H10
CAS Number:
Molecular Weight:
106.17
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing