Skip to Content
Merck
5-tert-Butyl-2-hydroxybenzaldehyde 98%

5-tert-Butyl-2-hydroxybenzaldehyde

5-tert-Butyl-2-hydroxybenzaldehyde 98%
Synonym(s):
5-tert-Butylsalicylaldehyde
Linear Formula:
(CH3)3CC6H3(OH)CHO
CAS Number:
Molecular Weight:
178.23

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing