Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
5-Sulfosalicylic acid dihydrate
Synonym(s):
5-Sulfosalicylic acid dihydrate, 2-Hydroxy-5-sulfobenzoic acid
Linear Formula:
HO
3
SC
6
H
3
-2-(OH)CO
2
H·2H
2
O
CAS Number:
5965-83-3
Molecular Weight:
254.21
EC Number:
202-555-6
Products
Product Category
organic acids (4)
building blocks (2)
Brand
Sigma-Aldrich (8)
Feature
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
ACS reagent (1)
Purum (1)
Purum P.A. (1)
Technique
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
8.00691
5-Sulfosalicylic acid dihydrate,
for synthesis
Expand
View Pricing
S2130
5-Sulfosalicylic acid dihydrate,
ReagentPlus
®
, ≥99%
Expand
View Pricing
247006
5-Sulfosalicylic acid dihydrate,
ACS reagent, ≥99%
Expand
View Pricing
28-5810
5-Sulfosalicylic acid dihydrate,
SAJ first grade, ≥99.0%
Expand
View Pricing
S3147
5-Sulfosalicylic acid dihydrate,
suitable for electrophoresis, ≥99%
Expand
View Pricing
S7422
5-Sulfosalicylic acid dihydrate,
BioXtra, ≥99.0%
Expand
View Pricing
86195
5-Sulfosalicylic acid dihydrate,
purum p.a., ≥98.0% (T)
Expand
View Pricing
28-5800
5-Sulfosalicylic acid dihydrate,
JIS special grade, ≥99.0
Expand
View Pricing