Skip to Content
AN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
4-Phenylphenol
Synonym(s):
4-Hydroxybiphenyl
Linear Formula:
C
6
H
5
C
6
H
4
OH
CAS Number:
92-69-3
Molecular Weight:
170.21
Products
Product Category
building blocks (2)
pesticides (1)
Brand
Sigma-Aldrich (2)
Boiling Point (°C)
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Product Line
Purity
Format
Quality Segment
Markush Class
Markush Group
Reference Material Type
Special Grade
certified reference material (1)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
134341
4-Phenylphenol,
97%
Expand
View Pricing
424625
4-Phenylphenol,
purified by sublimation, 99%
Expand
View Pricing
CRM69184
4-Phenylphenol,
certified reference material,
Trace
CERT
®
, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Expand
View Pricing