
4-Hydroxyisoleucine

Synonym(s):
4-Hydroxy-L-isoleucine, (2S,3R)-2-Amino-4-hydroxy-3-methylpentanoic acid
Empirical Formula (Hill Notation):
C6H13NO3
CAS Number:
Molecular Weight:
147.17
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing