Skip to Content
Merck
3-Methoxy-2-nitrobenzaldehyde ≥97%

3-Methoxy-2-nitrobenzaldehyde

3-Methoxy-2-nitrobenzaldehyde ≥97%
Linear Formula:
CH3OC6H3(NO2)CHO
CAS Number:
Molecular Weight:
181.15
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing