
2-(N-Ethylperfluorooctanesulfonamido)acetic acid

Synonym(s):
N-Ethyl-N-[(heptadecafluorooctyl)sulfonyl]glycine, N-Ethyl-perfluorooctanesulfonamidoacetic acid
Empirical Formula (Hill Notation):
C12H8F17NO4S
CAS Number:
Molecular Weight:
585.23
Products
Brand
Formula Weight
Manufacturer
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing