
2-Methyltetrahydrothiophen-3-one

Empirical Formula (Hill Notation):
C5H8OS
Products
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Purity
Agency
Quality Segment
Application
Organoleptic Class
Special Grade
Showing 1-1 of 1