Skip to Content
Merck
2-Bromoethanol analytical standard

2-Bromoethanol

2-Bromoethanol analytical standard
Synonym(s):
Ethylene bromohydrin
Linear Formula:
BrCH2CH2OH
CAS Number:
Molecular Weight:
124.96
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing