Skip to Content
Merck
2-Amino-5-chloropyridine British Pharmacopoeia (BP) Reference Standard

2-Amino-5-chloropyridine

2-Amino-5-chloropyridine British Pharmacopoeia (BP) Reference Standard
Empirical Formula (Hill Notation):
C5H5ClN2
CAS Number:
Molecular Weight:
128.56
EC Number:

Products

Product Category
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing