![2,2′-Bis[(4S)-4-benzyl-2-oxazoline] 98%](/deepweb/assets/sigmaaldrich/product/structures/139/783/42da3c77-52af-401b-8525-35d96415e284/640/42da3c77-52af-401b-8525-35d96415e284.png)
2,2′-Bis[(4S)-4-benzyl-2-oxazoline]
![2,2′-Bis[(4S)-4-benzyl-2-oxazoline] 98%](/deepweb/assets/sigmaaldrich/product/structures/139/783/42da3c77-52af-401b-8525-35d96415e284/640/42da3c77-52af-401b-8525-35d96415e284.png)
Synonym(s):
(S,S)-4,4′-Dibenzyl-2,2′-bi(2-oxazoline), (S,S)-2,2′-Bis(4-benzyl-2-oxazoline)
Empirical Formula (Hill Notation):
C20H20N2O2
CAS Number:
Molecular Weight:
320.39
Products
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing