
1,3,5-Tri-tert-butylbenzene

Linear Formula:
C6H3[C(CH3)3]3
CAS Number:
Molecular Weight:
246.43
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Reference Material Type
Special Grade