
1,2-Dimyristoyl-sn-glycero-3-phospho-rac-(1-glycerol) sodium salt

Synonym(s):
1,2-Ditetradecanoyl-sn-glycero-3-phospho-(1′-rac-glycerol) sodium salt, L-α-Phosphatidyl-DL-glycerol, dimyristoyl, L-α-Phosphatidyl-DL-glycerol, dimyristoyl sodium salt, DMPG, DMPG-Na, PG(14:0/14:0), Tetradecanoic acid 1-[[[(2,3-dihydroxypropoxy)hydroxyphosphinyl]oxy]methyl]-1,2-ethanediyl ester monosodium salt
Linear Formula:
C34H66O10PNa
CAS Number:
Molecular Weight:
688.85
Products
Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Optical Isomer
Showing 1-2 of 2