Skip to Content
Merck
All Photos(1)

Key Documents

1719532

USP

Withanoside IV

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(1α,3β,22R)-3-[(6-O-β-D-Glucopyranosyl-β-D-glucopyranosyl)oxy]-1,22,27-trihydroxy-ergosta-5,24-dien-26-oic acid δ-lactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H62O15
CAS Number:
Molecular Weight:
782.91
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

withania somnifera

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

C[C@H]([C@H]1CC(C)=C(CO)C(=O)O1)[C@H]2CC[C@H]3[C@@H]4CC=C5C[C@H](C[C@H](O)[C@]5(C)[C@H]4CC[C@]23C)O[C@@H]6O[C@H](CO[C@@H]7O[C@H](CO)[C@@H](O)[C@H](O)[C@H]7O)[C@@H](O)[C@H](O)[C@H]6O

InChI

1S/C40H62O15/c1-17-11-26(53-36(50)22(17)14-41)18(2)23-7-8-24-21-6-5-19-12-20(13-29(43)40(19,4)25(21)9-10-39(23,24)3)52-38-35(49)33(47)31(45)28(55-38)16-51-37-34(48)32(46)30(44)27(15-42)54-37/h5,18,20-21,23-35,37-38,41-49H,6-16H2,1-4H3/t18-,20+,21-,23+,24-,25-,26+,27+,28+,29-,30+,31+,32-,33-,34+,35+,37+,38+,39+,40-/m0/s1

InChI key

VUQQGHSDHGOYRH-IFUSOADVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Withanoside IV USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Ashwagandha Root
  • Ashwagandha Root Dry Extract
  • Ashwagandha Root Powder

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service