Key Documents
57633
Rotor Seal for Model 7125/7725/7725i Injector
Tefzel®
About This Item
UNSPSC Code:
41115854
material
Tefzel®
General description
Rotor Seal for Model 7125/7725/7725i Injector is an accessory for use with mainly Rheodyne® model 7725 and 7725i injectors for operation at pH 0-14.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
Rheodyne is a registered trademark of IDEX Health & Science, LLC
Tefzel is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)