Skip to Content
Merck
All Photos(1)

Key Documents

52776-U

Supelco

SupelMIP® Patulin solid phase extraction (SPE) Tube

Synonym(s):

MIP SPE (molecular imprinted) patulin, cartridge tube

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115712
NACRES:
NB.21

product name

SupelMIP® SPE - Patulin,

material

polypropylene tube

Quality Level

product line

SupelMIP®

composition

bed wt, 100 mg

packaging

pk of 50 ea

technique(s)

solid phase extraction (SPE): suitable

volume

3 mL

application(s)

food and beverages

General description

The SupelMIP SPE line consist s of highly cross-linked polymers that are engineered to extract a single analyte of interest or a class of structurally related analytes with an extremely high degree of selectivity. This is possible because selectivity is introduced during MIP synthesis in which a template molecule, designed to mimic the analyte, guides the formation of specific cavities or imprints that are sterically and chemically complementary to the analyte(s) of interest.

By careful design of the imprinting site, either by molecular modeling, experimental design, or screening methods, the binding cavities can be engineered to offer multiple interaction points with the analyte(s) of interest. This leads to a stronger interaction between the solid-phase and the analyte(s). As a consequence, harsher wash conditions can be tolerated during SPE methodology resulting in cleaner extracts. Because extraction selectivity is significantly improved, lower background is observed allowing analysts to achieve lower limits of detection.

Visit www.sigma-aldrich.com/supelmip to learn more.

Application

Developed for the class-selective extraction of the mycotoxin patulin from fruit, fruit products, and related sample matrices.

Features and Benefits

  • Highly selective SPE for trace analysis in complex matrices
  • Achieve lower detection limits
  • Significant time and cost savings
  • Improved MS-compatibility

Legal Information

SupelMIP is a registered trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service